Invitrogen
Manufacturer Code:PA542406
Catalog # PIPA542406
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human ABCA13 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP binding cassette transporter A13; ATP-binding cassette sub-family A (ABC1) member 13; ATP-binding cassette sub-family A member 13; ATP-binding cassette sub-family A member 13 variant 2; ATP-binding cassette sub-family A member 13 variant 3; ATP-binding cassette, sub-family A (ABC1), member 13
Gene Aliases: ABCA13
UniProt ID: (Human) Q86UQ4
Entrez Gene ID: (Human) 154664
Molecular Function:
ATP-binding cassette (ABC) transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.