Novus Biologicals
Manufacturer Code:NBP257346
Catalog # NBP257346
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DWLPLNQTFSQVSELVLNVTISTLTFLQQHGVAVTEPVYHLSMQNIVWDPQKVQYDLKSQFGFDDLHTEQILNSSAELKEIPTDTSLEKMVCSV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP binding cassette transporter A13; ATP binding cassette transporter A13 ATP-binding cassette sub-family A member 13 ATP-binding cassette sub-family A (ABC1) member 13 DKFZp313D2411 EC 2.7.11.25 EC 3.6.3 FLJ16398 FLJ33876 FLJ33951; ATP-binding cassette sub-family A member 13; ATP-binding cassette sub-family A member 13 variant 2; ATP-binding cassette sub-family A member 13 variant 3; ATP-binding cassette, sub-family A (ABC1), member 13
Gene Aliases: ABCA13
UniProt ID: (Human) Q86UQ4
Entrez Gene ID: (Human) 154664
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.