Abnova
Manufacturer Code:H00026154K
Catalog # 89-202-115
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ABCA12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human ABCA12 peptide using ARM Technology.
For Research Use Only
Protein Aliases: ATP-binding cassette 12; ATP-binding cassette sub-family A member 12; ATP-binding cassette transporter 12; ATP-binding cassette, sub-family A (ABC1), member 12; DKFZp434G232 FLJ41584 ICR2B LI2
Gene Aliases: ABC12; ABCA12; ARCI4A; ARCI4B; ICR2B; LI2
UniProt ID: (Human) Q86UK0
Entrez Gene ID: (Human) 26154
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.