Invitrogen
Manufacturer Code:MA521013
Catalog # PIMA521013
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 8F4 |
Immunogen | ABCA10 (NP&lowbar525021 1 a.a. &bsim 80 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ATP-binding cassette A10; ATP-binding cassette sub-family A member 10; ATP-binding cassette, sub-family A (ABC1), member 10; FKHL6 FREAC2
Gene Aliases: ABCA10; EST698739
UniProt ID: (Human) Q8WWZ4
Entrez Gene ID: (Human) 10349
Molecular Function:
ATP-binding cassette (ABC) transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.