Novus Biologicals
Manufacturer Code:NBP2825440.1ML
Catalog # NB030473
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C terminal region of human ABCA10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-binding cassette A10; ATP-binding cassette A10 ATP-binding cassette sub-family A member 10 ATP-binding cassette sub-family A (ABC1) member 10 EC 3.6.3 EC 3.6.3.41 EST698739; ATP-binding cassette sub-family A member 10; ATP-binding cassette, sub-family A (ABC1), member 10
Gene Aliases: ABCA10; EST698739
UniProt ID: (Human) Q8WWZ4
Entrez Gene ID: (Human) 10349
Molecular Function:
ATP-binding cassette (ABC) transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.