Abnova
Manufacturer Code:MAB8772
Catalog # 89-113-577
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AB.H10 |
Immunogen | A synthetic peptide corresponding to amino acids 1800-2260 at C-terminus end of human ABCA1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against synthetic peptide of ABCA1.
For Research Use Only
Protein Aliases: ABC-1; ABC1; ATP-binding cassette sub-family A member 1; ATP-binding cassette transporter 1; ATP-binding cassette transporter A1; ATP-binding cassette, sub-family A (ABC1), member 1; CERP; Cholesterol efflux regulatory protein; FLJ14958; HDLDT1; membrane-bound; MGC164864; MGC165011; Phospholipid-transporting ATPase ABCA1; TGD
Gene Aliases: ABC-1; ABC1; ABCA1; CERP; HDLDT1; TGD
UniProt ID: (Human) O95477
Entrez Gene ID: (Human) 19
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.