Invitrogen
Manufacturer Code:PA569245
Catalog # PIPA569245
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse ABCA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC-1; ABC-1 ABC1 CERP HDLDT1 TGD ATP-binding cassette sub-family A member 1 ATP-binding cassette transporter 1 ATP-binding cassette transporter A1 cholesterol efflux regulatory protein membrane-bound; ABCA1; ATP-binding cassette 1; ATP-binding cassette sub-family A member 1; ATP-binding cassette transporter 1; ATP-binding cassette transporter A1; ATP-binding cassette, sub-family A (ABC1), member 1; ATP-binding cassette, sub-family A member 1; Cholesterol efflux regulatory protein; membrane-bound; Phospholipid-transporting ATPase ABCA1
Gene Aliases: ABC-1; ABC1; ABCA1; CERP; HDLDT1; TGD
UniProt ID: (Human) O95477, (Mouse) P41233
Entrez Gene ID: (Horse) 100054241, (Dog) 481651, (Rabbit) 100356765, (Human) 19, (Bovine) 535379, (Pig) 100152112, (Mouse) 11303, (Rat) 313210
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.