R&D Systems
Manufacturer Code:AF7207
Catalog # AF7207
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human ABCA1 Phe1248-Gln1350 Accession # O95477 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC-1; ABC1 ABC-1 ABC1ATP-binding cassette transporter A1 ATP-binding cassette 1 ATP-binding cassette transporter 1 ATP-binding cassette sub-family A (ABC1) member 1 CERP Cholesterol efflux regulatory protein EC 2.7.7.8 EC 2.8.1.8 EC 3.6.3 EC 3.6.3.41 FLJ14958 HDLDT1 membrane-bound MGC164864 MGC165011 TGD TGDATP-binding cassette sub-family A member 1; ATP-binding cassette sub-family A member 1; ATP-binding cassette transporter 1; ATP-binding cassette transporter A1; ATP-binding cassette, sub-family A (ABC1), member 1; Cholesterol efflux regulatory protein; membrane-bound; Phospholipid-transporting ATPase ABCA1
Gene Aliases: ABC-1; ABC1; ABCA1; CERP; HDLDT1; TGD
UniProt ID: (Human) O95477
Entrez Gene ID: (Human) 19
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.