Invitrogen
Manufacturer Code:MA526824
Catalog # PIMA526824
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7H1 |
Immunogen | Human recombit protein fragment corresponding to amino acids 29-323 of ABAT produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: (S) 3 amino 2 methylpropionate transaminase (S)-3-amino-2-methylpropionate transaminase 4 aminobutyrate aminotransferase 4 aminobutyrate aminotransferase mitochondrial 4-aminobutyrate aminotransferase ABAT FLJ17813 FLJ30272 GABA aminotransferase GABA AT GABA T GABA transaminase GABA transferase GABA-AT GABA-T GABAT GABT&lowbarHUMAN Gamma amino N butyrate transaminase Gamma-amino-N-butyrate transaminase hCG1984265 L AIBAT L-AIBAT LAIBAT mitochondrial NPD009; (S)-3-amino-2-methylpropionate transaminase; 4-aminobutyrate aminotransferase, mitochondrial; 4-aminobutyrate transaminase; GABA aminotransferase; GABA transaminase; GABA transferase; GABA-AT; Gamma-amino-N-butyrate transaminase; L-AIBAT
Gene Aliases: ABAT; GABA-AT; GABAT; NPD009
UniProt ID: (Human) P80404
Entrez Gene ID: (Human) 18
Molecular Function:
transaminase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.