Invitrogen
Manufacturer Code:PA527405
Catalog # PIPA527405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 302 and 500 of Human ABAT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (S)-3-amino-2-methylpropionate transaminase; 4-aminobutyrate aminotransferase, brain isoform; 4-aminobutyrate aminotransferase, liver isoform; 4-aminobutyrate aminotransferase, mitochondrial; 4-aminobutyrate transaminase; beta-alanine oxoglutarate aminotransferase; GABA aminotransferase; GABA transaminase; GABA transferase; GABA-AT; GABA-AT GABAT NPD009 (S)-3-amino-2-methylpropionate transaminase 4-aminobutyrate aminotransferase mitochondrial 4-aminobutyrate transaminase GABA aminotransferase GABA transaminase GABA transferase gamma-amino-N-butyrate transaminase; GABA-T; Gamma-amino-N-butyrate transaminase; L-AIBAT
Gene Aliases: 9630038C02Rik; ABAT; AI255750; beta-AlaAT; ENSMUSG00000051226; GABA-AT; Gabaat; GABAT; Gm9851; I54; Laibat; NPD009; X61497
UniProt ID: (Human) P80404, (Rat) P50554, (Mouse) P61922
Entrez Gene ID: (Human) 18, (Rat) 81632, (Mouse) 268860
Molecular Function: transaminase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.