Novus Biologicals
Manufacturer Code:NBP133066
Catalog # NBP133066
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of human ABAT. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (S)-3-amino-2-methylpropionate transaminase; (S)-3-amino-2-methylpropionate transaminase 4-aminobutyrate aminotransferase 4-aminobutyrate aminotransferase mitochondrial 4-aminobutyrate transaminase EC 2.6.1.19 EC 2.6.1.22 FLJ17813 FLJ30272 G GABA aminotransferase GABA transaminase GABA transferase GABA-AT GABA-T GABATNPD009 Gamma-amino-N-butyrate transaminase hCG1984265 L-AIBAT; 4-aminobutyrate aminotransferase, mitochondrial; 4-aminobutyrate transaminase; GABA aminotransferase; GABA transaminase; GABA transferase; GABA-AT; Gamma-amino-N-butyrate transaminase; L-AIBAT
Gene Aliases: ABAT; GABA-AT; GABAT; NPD009
UniProt ID: (Human) P80404
Entrez Gene ID: (Human) 18
Molecular Function: transaminase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.