R&D Systems
Manufacturer Code:IC4866U100UG
Catalog # IC4866U100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 456006 |
Immunogen | <EM>E. coli</EM>-derived recombinant human AATK Asn421-Ala589 Accession # AAH47378 |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: AATYK; AATYK AATYKp35-binding protein apoptosis-associated tyrosine kinaseCDK5-binding protein Brain apoptosis-associated tyrosine kinase EC 2.7.11 KIAA0641p35BP Lemur tyrosine kinase 1 LMR1 LMR1EC 2.7.11.1 LMTK1 LMTK1AATYK1 p35BP serine/threonine-protein kinase LMTK1; Apoptosis-associated tyrosine kinase; Brain apoptosis-associated tyrosine kinase; CDK5-binding protein; Lemur tyrosine kinase 1; p35-binding protein; p35BP; protein phosphatase 1, regulatory subunit 77; Serine/threonine-protein kinase LMTK1
Gene Aliases: AATK; AATYK; AATYK1; KIAA0641; LMR1; LMTK1; p35BP; PPP1R77
UniProt ID: (Human) Q6ZMQ8
Entrez Gene ID: (Human) 9625
Molecular Function:
kinase
protein kinase
protein kinase receptor
receptor
transferase
tyrosine protein kinase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.