Invitrogen
Manufacturer Code:MA537670
Catalog # PIMA537670
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1459CT852.80.10 |
Immunogen | recombinant protein AATK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AATK AATYK aatyk1 apoptosis associated tyrosine kinase apoptosis-associated tyrosine kinase apoptosis-associated tyrosine kinase 1B brain apoptosis-associated tyrosine kinase CDK5-binding protein KIAA0641 Lemur tyrosine kinase 1 Lmr1 Lmtk1 mKIAA0641 p35-binding protein p35BP PPP1R77 protein phosphatase 1 regulatory subunit 77 serine/threonine-protein kinase LMTK1; AATYK; Apoptosis-associated tyrosine kinase; apoptosis-associated tyrosine kinase 1B; Brain apoptosis-associated tyrosine kinase; CDK5-binding protein; Lemur tyrosine kinase 1; p35-binding protein; p35BP; protein phosphatase 1, regulatory subunit 77; Serine/threonine-protein kinase LMTK1
Gene Aliases: AATK; AATYK; AATYK1; KIAA0641; LMR1; LMTK1; mKIAA0641; p35BP; PPP1R77
UniProt ID: (Human) Q6ZMQ8, (Mouse) Q80YE4
Entrez Gene ID: (Human) 9625, (Mouse) 11302
Molecular Function:
kinase
protein kinase
protein kinase receptor
receptor
transferase
tyrosine protein kinase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.