Novus Biologicals
Manufacturer Code:NBP29209320UL
Catalog # NB026079
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 180-400 of human AASS (NP_005754.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha-aminoadipate semialdehyde synthase; Alpha-aminoadipic semialdehyde synthase, mitochondrial; aminoadipate-semialdehyde synthase aminoadipic semialdehyde synthase LKR/SDH LKRSDH LORSDH lysine-2-oxoglutarate reductase mitochondrial; aminoadipic semialdehyde synthase; LKR; LKR/SDH; Lysine ketoglutarate reductase; lysine-2-oxoglutarate reductase; lysine-ketoglutarate reductase /saccharopine dehydrogenase; Saccharopine dehydrogenase; SDH
Gene Aliases: AASS; LKR/SDH; LKRSDH; LORSDH
UniProt ID: (Human) Q9UDR5
Entrez Gene ID: (Human) 10157
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.