Invitrogen
Manufacturer Code:PA543218
Catalog # PIPA543218
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human AASDHPPT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4'-phosphopantetheinyl transferase; AASD-PPT; Alpha-aminoadipic semialdehyde dehydrogenase-phosphopantetheinyl transferase; Aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase AASD-PPT CGI-80 DKFZp566E2346 LYS2 LYS5; holo ACP synthase; holo-[acyl-carrier-protein] synthase; L-aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase; LYS5 ortholog
Gene Aliases: AASD-PPT; AASDHPPT; ACPS; CGI-80; HAH-P; HSPC223; LYS2; LYS5; x0005
UniProt ID: (Human) Q9NRN7, (Rat) B2RYJ4
Entrez Gene ID: (Pig) 100525064, (Bovine) 519816, (Human) 60496, (Horse) 100061627, (Dog) 489426, (Rabbit) 100357546, (Rat) 300328, (Guinea pig) 100731306
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.