Proteintech
Manufacturer Code:112441AP150UL
Catalog # 50-172-6998
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AASDHPPT Fusion Protein Ag1763 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4'-phosphopantetheinyl transferase; AASD PPT AASDHPPT CGI 80 HAH P HSPC223 LYS2 LYS5 LYS5 ortholog x0005; AASD-PPT; Alpha-aminoadipic semialdehyde dehydrogenase-phosphopantetheinyl transferase; holo ACP synthase; holo-[acyl-carrier-protein] synthase; L-aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase; LYS5 ortholog
Gene Aliases: 2010309J24Rik; 2810407B07Rik; AASD-PPT; AASDHPPT; ACPS; CGI-80; HAH-P; HSPC223; LYS2; LYS5; x0005
UniProt ID: (Human) Q9NRN7, (Rat) B2RYJ4, (Mouse) Q9CQF6
Entrez Gene ID: (Human) 60496, (Rat) 300328, (Mouse) 67618
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.