Abnova
Manufacturer Code:H00132949K
Catalog # 89-206-784
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human AASDH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human AASDH peptide using ARM Technology.
For Research Use Only
Protein Aliases: 2-aminoadipic 6-semialdehyde dehydrogenase; ACSF4 LYS2 NRPS1098 NRPS998; Acyl-CoA synthetase family member 4; Beta-alanine-activating enzyme; non-ribosomal peptide synthetase 1098; non-ribosomal peptide synthetase 998; Protein NRPS998
Gene Aliases: AASDH; ACSF4; HSPC318; LYS2; NRPS1098; NRPS998; U26
UniProt ID: (Human) Q4L235
Entrez Gene ID: (Human) 132949
Molecular Function:
dehydrogenase
ligase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.