Novus Biologicals
Manufacturer Code:NBP292449100UL
Catalog # NB026768
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-309 of human AASDHPPT (NP_056238.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4&prime-phosphopantetheinyl transferase AASD-PPTDKFZp566E2346 Alpha-aminoadipic semialdehyde dehydrogenase-phosphopantetheinyl transferase aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase CGI-80 EC 2.7.8 EC 2.7.8.- L-aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase LYS2 LYS5 LYS5 ortholog; 4'-phosphopantetheinyl transferase; AASD-PPT; Alpha-aminoadipic semialdehyde dehydrogenase-phosphopantetheinyl transferase; holo ACP synthase; holo-[acyl-carrier-protein] synthase; L-aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase; LYS5 ortholog
Gene Aliases: AASD-PPT; AASDHPPT; ACPS; CGI-80; HAH-P; HSPC223; LYS2; LYS5; x0005
UniProt ID: (Human) Q9NRN7
Entrez Gene ID: (Human) 60496
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.