Novus Biologicals
Manufacturer Code:NBP290276AF405
Catalog # NB112075
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 12 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human AARSD1 (Q9BTE6-2 Thr176-Glu525). |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Alanyl-tRNA editing protein Aarsd1; alanyl-tRNA editing protein Aarsd1 alanyl-tRNA synthetase domain containing 1 Alanyl-tRNA synthetase domain-containing protein 1 MGC2744; Alanyl-tRNA synthetase domain-containing protein 1
Gene Aliases: AARSD1
UniProt ID: (Human) Q9BTE6
Entrez Gene ID: (Human) 80755
Molecular Function: RNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.