Novus Biologicals
Manufacturer Code:NBP226119
Catalog # NBP226119
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-QHRARQAEPVQK corresponding to internal region according to NP_065796.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: AARS2; alanine tRNA ligase 2 mitochondrial (putative) alanine--tRNA ligase alanyl-tRNA synthetase 2 mitochondrial (putative) alanyl-tRNA synthetase mitochondrial alaRS bA444E17.1 COXPD8 KIAA1270 MTALARS MT-ALARS probable alanyl-tRNA synthetase mitochondrial; alanine tRNA ligase 2, mitochondrial (putative); Alanine--tRNA ligase, mitochondrial; Alanyl-tRNA synthetase; alanyl-tRNA synthetase 2, mitochondrial (putative); alanyl-tRNA synthetase like; AlaRS; probable alanyl-tRNA synthetase, mitochondrial
Gene Aliases: AARS2; AARSL; COXPD8; KIAA1270; LKENP; MT-ALARS; MTALARS
UniProt ID: (Human) Q5JTZ9
Entrez Gene ID: (Human) 57505
Molecular Function:
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.