Novus Biologicals
Manufacturer Code:NBP2825430.1ML
Catalog # NB030472
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human AAR2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AAR2 splicing factor homolog; AAR2 splicing factor homolog (S. cerevisiae) bA234K24.2 C20orf4 CGI-23 chromosome 20 open reading frame 4 DKFZp564N1363 hypothetical protein LOC25980; Protein AAR2 homolog
Gene Aliases: AAR2; C20orf4; CGI-23; PRO0225
UniProt ID: (Human) Q9Y312
Entrez Gene ID: (Human) 25980
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.