Abnova
Manufacturer Code:PAB12505
Catalog # 89-115-879
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Human AANAT. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for short term. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against AANAT.
For Research Use Only
Protein Aliases: AA-NAT; Aralkylamine N-acetyltransferase; arylalkylamine N-acetyltransferase; Serotonin acetylase; Serotonin N-acetyltransferase; SNAT
Gene Aliases: AANAT; DSPS; SNAT
UniProt ID: (Human) Q16613
Entrez Gene ID: (Human) 15
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.