Novus Biologicals
Manufacturer Code:NBP271840AF700
Catalog # N271840A700
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C4 |
Immunogen | Full length recombinant protein of human C11orf67 (NP_078960) produced in E.coli. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: adipogenesis associated Mth938 domain containing C11orf67 chromosome 11 open reading frame 67 CK067 FLJ21035 hypothetical protein LOC28971 MGC3367 PTD015; Adipogenesis associated Mth938 domain-containing protein; adipogenesis associated, Mth938 domain containing; Mth938 domain-containing protein; UPF0366 protein C11orf67
Gene Aliases: AAMDC; C11orf67; CK067; PTD015
UniProt ID: (Human) Q9H7C9
Entrez Gene ID: (Human) 28971
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.