Invitrogen
Manufacturer Code:PA519202
Catalog # PIPA519202
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (GSPRTSQQNVYNPSE) corresponding to the internal amino acids of AAK1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: AAK1 AP2 associated kinase 1 DKFZp686F03202 DKFZp686K16132 FLJ23712 FLJ25931 FLJ31060 FLJ42882 FLJ45252 KIAA1048 MGC138170 MGC164568 MGC164570 adaptor-associated kinase 1; Adaptor-associated kinase 1; AP2-associated protein kinase 1
Gene Aliases: AAK1; KIAA1048
UniProt ID: (Human) Q2M2I8
Entrez Gene ID: (Human) 22848
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.