Invitrogen
Manufacturer Code:PA5114797
Catalog # PIPA5114797
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human AAK1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5530400K14Rik 9630042K20 Aak1 adaptor-associated kinase 1 AP2 associated kinase 1 AP2-associated protein kinase 1 AU067724 AU067726 BC028270 C79663 D6Ertd245e DKFZp686F03202 DKFZp686K16132 FLJ23712 FLJ25931 FLJ31060 FLJ42882 FLJ45252 KIAA1048 MGC138170 MGC164568 MGC164570 mKIAA1048 R75501 RGD1563580; Adaptor-associated kinase 1; AP2 associated kinase 1; AP2-associated protein kinase 1; Uncharacterized protein FLJ45252 homolog
Gene Aliases: 5530400K14Rik; 9630042K20; AAK1; AU067724; AU067726; BC028270; C79663; D6Ertd245e; KIAA1048; mKIAA1048; R75501; RGD1563580
UniProt ID: (Human) Q2M2I8, (Mouse) Q6PIU9
Entrez Gene ID: (Human) 22848, (Mouse) 269774, (Rat) 500244
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.