Proteintech
Manufacturer Code:273191AP150UL
Catalog # 50-173-5640
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AAGAB Fusion Protein Ag26340 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AAGAB FLJ11506; Alpha- and gamma-adaptin-binding protein p34
Gene Aliases: 2310007F21Rik; AAGAB; KPPP1; p34; PPKP1; PPKP1A
UniProt ID: (Human) Q6PD74, (Mouse) Q8R2R3
Entrez Gene ID: (Human) 79719, (Mouse) 66939
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.