Novus Biologicals
Manufacturer Code:NBP276805
Catalog # NBP276805
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-RTRNSYIKWLDQN from the C Terminus of the protein sequence according to NP_065843.3 NP_001139748.2 NP_001139749.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
Protein Aliases: AADACL1 EC 3.1.1.79 KIAA1363EC 3.1.1 NCEHArylacetamide deacetylase-like 1EC 3.1.1.- neutral cholesterol ester hydrolase 1; Arylacetamide deacetylase-like 1; NCEH; Neutral cholesterol ester hydrolase 1
Gene Aliases: AADACL1; KIAA1363; NCEH; NCEH1
UniProt ID: (Human) Q6PIU2
Entrez Gene ID: (Human) 57552
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.