Novus Biologicals
Manufacturer Code:NBP271358F
Catalog # NBP271358F
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5B10 |
Immunogen | Recombinant protein fragment corresponding to amino acids 322-546 of human AAAS (NP_056480) produced in E.coli. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: AAA AAASb achalasia adrenocortical insufficiency alacrimia achalasia adrenocortical insufficiency alacrimia (Allgrove triple-A) ADRACALA Adracalin ALADIN Allgrove triple-A DKFZp586G1624; achalasia, adrenocortical insufficiency, alacrimia; Adracalin; Aladin; Allgrove, triple-A
Gene Aliases: AAA; AAAS; AAASb; ADRACALA; ADRACALIN; ALADIN; GL003
UniProt ID: (Human) Q9NRG9
Entrez Gene ID: (Human) 8086
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.