OriGene
Manufacturer Code:TA800401
Catalog # 50-167-8297
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1F7 |
Immunogen | Human recombit protein fragment corresponding to amino acids 121-340 of human A4GNT produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Alpha-1,4-N-acetylglucosaminyltransferase; Alpha4GnT
Gene Aliases: A4GNT; Alpha4gnt; AV080780; Gm798
UniProt ID: (Human) Q9UNA3, (Mouse) Q14BT6
Entrez Gene ID: (Dog) 485683, (Human) 51146, (Rat) 685758, (Mouse) 333424
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.