R&D Systems
Manufacturer Code:AF8555
Catalog # AF8555
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human alpha-14-N-Acetylglucosaminyltransferase 4/A4GNT Leu25-Lys340 Accession # Q9UNA3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 12 months from date of receipt -20 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after opening. 6 months -20 degreesC under sterile conditions after opening. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-1,4-N-acetylglucosaminyltransferase; alpha-14-N-acetylglucosaminyltransferase alpha4GnT; Alpha4GnT
Gene Aliases: A4GNT; alpha4GnT
UniProt ID: (Human) Q9UNA3
Entrez Gene ID: (Human) 51146
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.