Proteintech
Manufacturer Code:123921AP150UL
Catalog # 50-172-7951
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A4GALT Fusion Protein Ag3069 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A4galt; A4GALT A4GALT1 CD77 synthase Gb3 synthase; alpha 14-galactosyltransferase; Alpha-1,4-galactosyltransferase; Alpha-1,4-N-acetylglucosaminyltransferase; Alpha4Gal-T1; CD77 synthase; GB3 synthase; Gb3/CD77 synthase; Globotriaosylceramide synthase; Lactosylceramide 4-alpha-galactosyltransferase; P blood group (P one antigen); P(k) antigen synthase; P1/Pk synthase; truncated alpha 1,4-galactosyltransferase; UDP-galactose:beta-D-galactosyl-beta1-R 4-alpha-D-galactosyltransferase
Gene Aliases: A14GALT; A4GALT; A4GALT1; Gb3S; P(k); P1; P1PK; PK
UniProt ID: (Human) Q9NPC4, (Mouse) Q67BJ4
Entrez Gene ID: (Human) 53947, (Mouse) 239559
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.