Invitrogen
Manufacturer Code:PA587197
Catalog # PIPA587197
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human GPA33 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A33; A33 antigen; Cell surface A33 antigen; Glycoprotein A33; glycoprotein A33 (transmembrane); mA33; transmembrane glycoprotein A33
Gene Aliases: 2010310L10Rik; 2210401D16Rik; A33; BB116197; GPA33
UniProt ID: (Human) Q99795, (Mouse) Q9JKA5
Entrez Gene ID: (Human) 10223, (Mouse) 59290, (Rat) 360873
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.