Invitrogen
Manufacturer Code:PA553866
Catalog # PIPA553866
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human A33 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell surface A33 antigen; Glycoprotein A33; glycoprotein A33 (transmembrane); GPA33 A33 cell surface A33 antigen Glycoprotein A33 glycoprotein A33 (transmembrane) MGC129986 MGC129987 transmembrane glycoprotein A33; transmembrane glycoprotein A33
Gene Aliases: A33; GPA33
UniProt ID: (Human) Q99795
Entrez Gene ID: (Human) 10223
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.