Invitrogen
Manufacturer Code:PA5115314
Catalog # PIPA5115314
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human GPA33 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2010310L10Rik 2210401D16Rik A33 A33 antigen BB116197 Cell surface A33 antigen Glycoprotein A33 glycoprotein A33 (transmembrane) GPA33 mA33 MGC129986 MGC129987 transmembrane glycoprotein A33; A33 antigen; Cell surface A33 antigen; Glycoprotein A33; glycoprotein A33 (transmembrane); mA33; transmembrane glycoprotein A33
Gene Aliases: 2010310L10Rik; 2210401D16Rik; A33; BB116197; GPA33
UniProt ID: (Human) Q99795, (Mouse) Q9JKA5
Entrez Gene ID: (Human) 10223, (Mouse) 59290
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.