Invitrogen
Manufacturer Code:PA524660
Catalog # PIPA524660
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 345-373 amino acids from the N-terminal region of human A2ML1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha-2-macroglobulin-like 1; Alpha-2-macroglobulin-like protein 1; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 9; C3 and PZP-like, alpha-2-macroglobulin domain containing 9; CPAMD9 C3 and PZP-like alpha-2-macroglobulin domain containing 9 alpha-2-macroglobulin-like protein 1; Ovostatin homolog
Gene Aliases: A2ML1; CPAMD9; Ovos
UniProt ID: (Human) A8K2U0, (Mouse) Q3UU35
Entrez Gene ID: (Human) 144568, (Mouse) 232400
Molecular Function:
complement component
cytokine
defense/immunity protein
enzyme modulator
protease inhibitor
serine protease inhibitor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.