Invitrogen
Manufacturer Code:PA5102427
Catalog # PIPA5102427
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human A2M corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A2M A2m1 A2maa A2MAC1 A2MD A2mp Alpha 2 M alpha-2-M Alpha-2-macroglobulin alpha-2-macroglobulin-P C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5 CPAMD5 DKFZp779B086 FWP007 Mam S863 7 S863-7; A2mp; Alpha-2-M; Alpha-2-macroglobulin; Alpha-2-macroglobulin-P; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5
Gene Aliases: A2M; A2MD; A2mp; CPAMD5; FWP007; S863-7
UniProt ID: (Human) P01023, (Mouse) Q6GQT1
Entrez Gene ID: (Human) 2, (Mouse) 232345
Molecular Function:
complement component
cytokine
defense/immunity protein
enzyme modulator
protease inhibitor
serine protease inhibitor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.