Abnova
Manufacturer Code:PAB29697
Catalog # 89-933-493
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Native purified rat A2m. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against rat A2m.
For Research Use Only
Protein Aliases: A2m1; A2maa; A2MAC1; Alpha-2-M; Alpha-2-macroglobulin; alpha-2-macroglobulin-P; Mam; MGC114358; RATA2MAC1
Gene Aliases: A2m; A2m1; A2maa; A2MAC1; Mam
UniProt ID: (Rat) P06238
Entrez Gene ID: (Rat) 24153
Molecular Function: complement component cytokine defense/immunity protein enzyme modulator protease inhibitor serine protease inhibitor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.