Invitrogen
Manufacturer Code:PA575795
Catalog # PIPA575795
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human A20A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ankyrin repeat domain 20 family, member A1; ankyrin repeat domain 20A; Ankyrin repeat domain-containing protein 20A1; Ankyrin repeat domain-containing protein 20A1 ANKRD20A1 ANKRD20A
Gene Aliases: ANKRD20A; ANKRD20A1
UniProt ID: (Human) Q5TYW2
Entrez Gene ID: (Human) 84210
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.