Novus Biologicals
Manufacturer Code:NBP177533SS
Catalog # NBP177533SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 59A426 |
Immunogen | Full length recombinant human A20/TNFAIP3 Antibody (59A426). The epitope has been mapped to the C-terminal portion of A20 (amino acids 440-790) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A20p37; A20p50; A20TNFA1P2 EC 3.4.19.12 EC 6.3.2.- MGC104522 MGC138687 OTU domain-containing protein 7C OTUD7CMGC138688 Putative DNA-binding protein A20 TNF alpha-induced protein 3 tumor necrosis factor alpha-induced protein 3 tumor necrosis factor inducible protein A20 tumor necrosis factor alpha-induced protein 3 Zinc finger protein A20; OTU domain-containing protein 7C; Putative DNA-binding protein A20; TNF alpha-induced protein 3; Tumor necrosis factor alpha-induced protein 3; tumor necrosis factor inducible protein A20; tumor necrosis factor, alpha induced protein 3; tumor necrosis factor, alpha-induced protein 3; Zinc finger protein A20
Gene Aliases: A20; AISBL; OTUD7C; TNFA1P2; TNFAIP3
UniProt ID: (Human) P21580
Entrez Gene ID: (Human) 7128
Molecular Function:
DNA binding protein
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.