Abnova
Manufacturer Code:MAB0794
Catalog # 89-108-139
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 54B12 |
Immunogen | Native purified human A1BG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against native A1BG.
For Research Use Only
Protein Aliases: A1B; ABG; Alpha-1-B glycoprotein; Alpha-1B-glycoprotein; DKFZp686F0970; epididymis secretory sperm binding protein Li 163pA; GAB; HEL-S-163pA; HYST2477
Gene Aliases: A1B; A1BG; ABG; GAB; HYST2477
UniProt ID: (Human) P04217
Entrez Gene ID: (Human) 1
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.