Novus Biologicals
Manufacturer Code:NBP147478
Catalog # NBP147478
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4B5 |
Immunogen | Purified recombinant fragment of human A1BG expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A1B ABG alpha-1-B glycoproteinGAB alpha-1B-glycoprotein DKFZp686F0970 HYST2477; Alpha-1-B glycoprotein; Alpha-1B-glycoprotein; epididymis secretory sperm binding protein Li 163pA; HEL-S-163pA
Gene Aliases: A1B; A1BG; ABG; GAB; HYST2477
UniProt ID: (Human) P04217
Entrez Gene ID: (Human) 1
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.