Invitrogen
Manufacturer Code:PA575479
Catalog # PIPA575479
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 182-227 of Human A1BG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A1bg {ECO:0000312|RGD:69417}; Alpha-1-B glycoprotein; Alpha-1B-glycoprotein; Alpha-1B-glycoprotein Alpha-1-B glycoprotein alpha-1B-glycoprotein alpha1B-glycoprotein alpha-1-B-glycoprotein; C44; epididymis secretory sperm binding protein Li 163pA; HEL-S-163pA; Liver regeneration-related protein 1
Gene Aliases: A1B; A1BG; ABG; C44; GAB; HYST2477
UniProt ID: (Human) P04217, (Rat) Q9EPH1, (Mouse) Q19LI2
Entrez Gene ID: (Human) 1, (Rat) 140656, (Mouse) 117586
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.