Abnova
Manufacturer Code:H00011214M10
Catalog # 89-001-211
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3D6 |
Immunogen | AKAP13 (NP_006729 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant AKAP13.
For Research Use Only
Protein Aliases: A kinase (PRKA) anchor protein 13; A-kinase anchor protein 13; A-kinase anchoring protein; AKAP-13; AKAP-Lbc; ARHGEF13; Breast cancer nuclear receptor-binding auxiliary protein; BRX; c-lbc; FLJ11952; FLJ43341; Guanine nucleotide exchange factor Lbc; HA-3; Ht31; Human thyroid-anchoring protein 31; LBC; LBC oncogene; Lymphoid blast crisis oncogene; Non-oncogenic Rho GTPase-specific GTP exchange factor; p47; PRKA13; Protein kinase A-anchoring protein 13; PROTO-LB; PROTO-LBC
Gene Aliases: AKAP-13; AKAP-Lbc; AKAP13; ARHGEF13; BRX; c-lbc; HA-3; HT31; LBC; p47; PRKA13; PROTO-LB; PROTO-LBC
UniProt ID: (Human) Q12802
Entrez Gene ID: (Human) 11214
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.