R&D Systems
Manufacturer Code:MAB6607
Catalog # MAB6607
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 684128 |
Immunogen | <em>E. coli</em>-derived recombinant human A-Raf Asn150-Lys314 Accession # P10398 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A-Raf proto-oncogene serine/threonine-protein kinase; ARaf ARAF1A-RAF EC 2.7.11 EC 2.7.11.1 Oncogene ARAF1 PKS PKS2 PKS2A-Raf proto-oncogene serine/threonine-protein kinase Proto-oncogene A-Raf Proto-oncogene A-Raf-1 Proto-oncogene Pks RAFA1 Ras-binding protein DA-Raf serine/threonine-protein kinase A-Raf v-raf murine sarcoma 3611 viral oncogene homolog v-raf murine sarcoma 3611 viral oncogene homolog 1; Oncogene ARAF1; Proto-oncogene A-Raf; Proto-oncogene A-Raf-1; Proto-oncogene Pks; Ras-binding protein DA-Raf; Serine/threonine-protein kinase A-Raf; v-raf murine sarcoma 3611 viral oncogene homolog 1; v-raf murine sarcoma 3611 viral oncogene-like protein
Gene Aliases: A-RAF; ARAF; ARAF1; PKS; PKS2; RAFA1
UniProt ID: (Human) P10398
Entrez Gene ID: (Human) 369
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.