Invitrogen
Manufacturer Code:MA121315D650
Catalog # MA121315D65
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | HIS.H8 |
Immunogen | 6x His synthetic peptide. |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: His Tag Histidine Tag Histidine Epitope Tag 6X His eptope tag 6His tag 6X His
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.