Abnova
Manufacturer Code:PAB15845
Catalog # 89-117-907
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant GST fusion protein corresponding to 181 amino acids of mouse 6430573F11Rik. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant 6430573F11Rik.
For Research Use Only
Protein Aliases: Probable tRNA methyltransferase 9-like protein; Probable tRNA methyltransferase 9B
Gene Aliases: Kiaa1456; TRM9L; Trmt9b; Trmt9l
UniProt ID: (Mouse) Q80WQ4
Entrez Gene ID: (Mouse) 319582
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.