Novus Biologicals
Manufacturer Code:NBP29244120UL
Catalog # NB026753
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human RPL23 (NP_000969.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 60S ribosomal protein L17; 60S ribosomal protein L23; Large ribosomal subunit protein uL14; MGC111167 MGC117346 MGC7200860S ribosomal protein L1760S ribosomal protein L23 ribosomal protein L17 ribosomal protein L23 rpL17; ribosomal protein L17
Gene Aliases: L23; rpL17; RPL23
UniProt ID: (Human) P62829
Entrez Gene ID: (Human) 9349
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.