Invitrogen
Manufacturer Code:PA567949
Catalog # PIPA567949
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human 5T4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5T4 oncofetal antigen; 5T4 oncofetal trophoblast glycoprotein; 5T4 oncotrophoblast glycoprotein; M6P1; M6P1 TPBG 5T4 oncofetal trophoblast glycoprotein 5T4 oncotrophoblast glycoprotein 5T4AG 5T4-AG 5T4M6P15T4 oncofetal antigen trophoblast glycoprotein; Trophoblast glycoprotein; WAIF1; Wnt-activated inhibitory factor 1
Gene Aliases: 5T4; 5T4AG; AW495680; M6P1; TPBG; WAIF1
UniProt ID: (Human) Q13641, (Mouse) Q9Z0L0
Entrez Gene ID: (Human) 7162, (Mouse) 21983
Molecular Function:
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.