Novus Biologicals
Manufacturer Code:NB600412SS
Catalog # NB600412SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 58K-9 |
Immunogen | 58K Golgi Protein purified from rat liver [UniProt# O88618] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Formimidoyltetrahydrofolate cyclodeaminase; formimidoyltransferase cyclodeaminase formimidoyltransferase-cyclodeaminase formiminotransferase cyclodeaminase Formiminotransferase-cyclodeaminase human formiminotransferase cyclodeaminase EC 4.3.1.410formiminotransferase-cyclodeaminase LCHC1; Formimidoyltransferase-cyclodeaminase; Formiminotetrahydrofolate cyclodeaminase; formiminotransferase cyclodeaminase; Formiminotransferase-cyclodeaminase; FTCD; Glutamate formimidoyltransferase; Glutamate formiminotransferase; Glutamate formyltransferase; LCHC1
Gene Aliases: FTCD; LCHC1
UniProt ID: (Human) O95954
Entrez Gene ID: (Human) 10841
Molecular Function:
deaminase
hydrolase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.