Novus Biologicals
Manufacturer Code:NB1002858
Catalog # NB1002858
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 425 and 475 of human Apoptosis-Stimulating Protein of p53 2 (53BP2) using the numbering given in entry NP_001026855.1 (GeneID 7159). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 53BP2; Apoptosis-stimulating of p53 protein 2; apoptosis-stimulating protein of p53 2 ASPP2P53BP253BP2apoptosis-stimulating of p53 protein 2 Bbp Bcl2-binding protein p53-binding protein 2 p53BP2 PPP1R13A Renal carcinoma antigen NY-REN-51 tumor protein p53 binding protein 2 tumor protein p53-binding protein 2 Tumor suppressor p53-binding protein 2; apoptosis-stimulating protein of p53, 2; Bbp; BCL2-binding protein; Renal carcinoma antigen NY-REN-51; tumor protein p53 binding protein, 2; Tumor suppressor p53-binding protein 2
Gene Aliases: 53BP2; ASPP2; BBP; P53BP2; PPP1R13A; TP53BP2
UniProt ID: (Human) Q13625
Entrez Gene ID: (Human) 7159
Molecular Function:
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.